About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Binding type:
non-treated surface
Material:
borosilicate glass 3.3
Sterility:
non-sterile
Packaging:
pkg of 12 ea
Feature:
autoclavable, graduated
material
borosilicate glass 3.3
joint
joint straight long neck
sterility
non-sterile
feature
autoclavable, graduated
packaging
pkg of 12 ea
manufacturer/tradename
DWK Life Sciences Kimble® 25615-2000
capacity
2 L
flask diam. × H
161 × 288
binding type
non-treated surface
General description
Microbiology is an important field of biology and medicine and deals with microorganisms (bacteria, fungi, protozoa or viruses) and their habitats and conditions. For this purpose, it is essential to produce pure culture media on which the corresponding microorganisms can grow. Irradiation of the media with IR or UV light can accelerate the growth of the cultures. Culture flasks have large bottom surfaces that are as flat as possible so that the layer thickness of the medium is uniformly high.
Application
Cell culture, microbiology, fermentation
Features and Benefits
- Complies with ASTM Specification E1404, Type IV, Class I requirements
- Manufactured from borosilicate glass 3.3
- KIMAX® flask has a thick-walled tubing neck with a plain fire-polished finish sealed to an Erlenmeyer-shaped body
- Autoclavable
- All flasks have durable white ceramic enamel scales to indicate approximate volumes at various levels
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Still not finding the right product?
Explore all of our products under KIMBLE® KIMAX® Cell Culture Flask
Choose from one of the most recent versions:
Already Own This Product?