Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41121818
NACRES:
NB.17
Material:
PETG
Sterility:
sterile; γ-irradiated
Feature:
RNase and DNase free, graduated, with cap (tamper evident)
Fitting:
(53B)
Packaging:
pkg of 6 ea
material
PETG
joint
joint threaded
sterility
sterile; γ-irradiated
feature
RNase and DNase free, graduated, with cap (tamper evident)
packaging
pkg of 6 ea
manufacturer/tradename
DWK Life Sciences WHEATON® WPBGC2000S
H × W
266 mm × 115 mm
capacity
2 L , 2000 mL
fitting
(53B)
General description
WHEATON® PETG Media Bottles are made of Eastman Tritan Eastar Copolyester PETG which is the resin of choice for single use labware and medical devices. This resin is Food Contact Status compliant. The PETG is an amorphous material that displays performance properties including clarity and toughness as well as excellent gas barrier and chemical inertness. The products can be safely sterilized with gamma irradiation without property loss or color shift. WHEATON® PETG Square / Octagonal Bottles are provided gamma irradiated sterilized to SAL 10-6 with blue caps attached. Each case is clean room packaged for sterile production with multi-barrier elastomer packaging. Available with tamper evident caps (see name).
Application
Storage, Media prep, Liquid Handling
Features and Benefits
- Transparent PETG
- Available in sizes from 30mL to 2L
- Choice of sterile standard screw cap or tamper-evident cap ensuring product integrity prior to use
- Leak free performance ensures suitability for transport and storage applications
- Square/octagonal footprint for efficient use of storage space
- DNase- and RNase-Free
- Moulded graduations provide volume guidelines
- Excellent gas barrier properties, ensuring pH stability
- Recommended temperature range +70°C to -40°CSterile and can be re-sterilized
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Still not finding the right product?
Explore all of our products under WHEATON® PETG Media Bottle
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.