Skip to Content
Merck

PLA0109

Rabbit anti-CRM1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CRM1, chromosome region maintenance 1 homolog, chromosome region maintenance 1 homolog (yeast), chromosome region maintenance 1 protein homolog, emb, exp1, exportin 1 (CRM1 homolog; yeast), exportin 1 (CRM1; yeast; homolog), exportin-1 (required for chromosome region maintenance)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:1,000-1:5,000, immunoprecipitation (IP): 2-10 μg/mg, western blot: 1:2,000-1:10,000

accession no.

NP_003391.1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... CRM1(7514)

Immunogen

The epitope recognized by PLA0109 maps to a region between residue 1025 and the C-terminus (residue 1071) of human Chromosome Region Maintenance 1 (Exportin 1) using the numbering given in entry NP_003391.1 (GeneID 7514).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Chromosome region maintenance 1 protein homolog (CRM1) is the Ran-GTP-binding nuclear export receptor for proteins bearing a leucine rich nuclear export signal and RNAs. CRM1 also possesses a mitotic function as it localizes to kinetochores, and with Ran-GTP, recruits Ran-GTPase-activating protein (Ran-GAP1) and Ran-binding protein (Ran-BP2) for proper mitotic spindle assembly and chromosome segregation. Additionally, CRM1 is suggested to function as a loading dock of various cell cycle and checkpoint factors such as cyclins, Cdks, p53 and BRCA1, to regulate proper chromosome duplication and genomic stability.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library