Skip to Content

RAB1046

Human FGF-21 ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum sample(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 8 pg/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGF21(26291)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

Fibroblast growth factor 21 (FGF-21) plays a vital role in the regulation of glucose uptake in mouse 3T3-L1 and primary human adipocytes. In addition, it is also implicated in lipid metabolism and in energy balance. FGF-21 plays a key role in controlling oxidative stress cell response. Overexpression of the gene has been associated with the development of acute pancreatitis, obesity, insulin resistance and metabolic syndrome. Additionally, elevated expression of FGF-21 has been observed in patients with insulin resistance and type 2 diabetes mellitus (T2DM). Thus, it can be considered as a potent target for an effective treatment of diabetes.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library