Skip to Content

SAB4702012

Monoclonal Anti-CD307b purified produced in mouse

Synonym(s):

FcRH2, FcRL2, IFGP4, IRTA4, SPAP1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
B24, monoclonal
Application:
FACS
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

unconjugated

Quality Segment

antibody product type

primary antibodies

clone

B24, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/ml

technique(s)

flow cytometry: suitable

isotype

IgG1κ

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FCRL2(79368)

General description

CD307b is a type I transmembrane glycoprotein of the Fc receptor family. It contains one ITAM motif and two ITIM motifs in its cytoplasmic domain. It is expressed in spleen and lymph nodes in mature B cells and memory B cells. CD307b may be a prognostic marker for chronic lymphocytic leukemia.

Immunogen

DNA-immunization followed by a boost with CD307b transfected cells

Biochem/physiol Actions

The mouse monoclonal antibody B24 recognizes an extracellular epitope of CD307b, a transmembrane glycoprotein expressed mainly in mature and memory B cells.

Physical form

Phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified by protein-A affinity chromatography.
Store at 2-8°C. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library