Skip to Content

SAB6010010

Anti-monomethyl-Histone H3.1(R17) antibody, clone 3E10, Rabbit Monoclonal

recombinant, expressed in HEK293F cells

Synonym(s):

Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l, HIST1H3A, H3FA, AND, HIST1H3B, H3FL, AND, HIST1H3C, H3FC, AND, HIST1H3D, H3FB, AND, HIST1H3E, H3FD, AND, HIST1H3F, H3FI, AND, HIST1H3G, H3FH, AND, HIST1H3H, H3FK, AND, HIST1H3I, H3FF, AND, HIST1H3J, H3FJ

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
3E10, recombinant monoclonal
Application:
ELISA, FACS, IF, IHC
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in HEK 293 F cells

Quality Segment

conjugate

unconjugated

antibody product type

primary antibodies

clone

3E10, recombinant monoclonal

form

liquid

mol wt

150 kDa

purified by

affinity chromatography

species reactivity

human

concentration

1.15mg/ml

technique(s)

ELISA: 2000:10000, flow cytometry: 1:50-1:200, immunofluorescence: 1:50-1:200, immunohistochemistry: 1:50-1:200

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

-10 to -25°C

target post-translational modification

monomethylation (Arg17)

General description

Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.

Immunogen

A synthesized peptide derived from human Mono-methyl-Histone H3.1 (R17)

Physical form

Rabbit IgG in phosphate buffered saline, pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Preparation Note

Upon receipt, store at -20°C or -80°C. Avoid repeated freeze.

Other Notes

AP-(Me)R-KQ


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library