Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
25 mg | Please contact Customer Service for Availability | $704.00 |
About This Item
Empirical Formula (Hill Notation):
C14H17F4NO2
Molecular Weight:
307.28
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
(Viscous Oil)
$704.00
Please contact Customer Service for Availability
Quality Segment
assay
≥98% (HPLC)
form
(Viscous Oil)
color
yellow to brown
storage temp.
2-8°C
SMILES string
O[C@]1([C@@H](CCCC1)NC2=CC=C(C(OC)=C2)F)C(F)(F)F
InChI
1S/C14H17F4NO2/c1-21-11-8-9(5-6-10(11)15)19-12-4-2-3-7-13(12,20)14(16,17)18/h5-6,8,12,19-20H,2-4,7H2,1H3/t12-,13-/m1/s1
InChI key
CWRARANSONEPBZ-CHWSQXEVSA-N
Biochem/physiol Actions
BAY-9897 is an inactive analog of and a negative control for the orally active, potent and selective TRPA1 inhibitor BAY-390 (human/rat IC50 >25 µM vs.16/63 nM using BAY-390; cellular Ca++ influx by FLIPR).
Inactive analog of and a negative control for the orally active, potent and selective TRPA1 inhibitor BAY-390.
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥95% (HPLC) | assay ≥98% (HPLC) |
| form (Viscous Oil) | form powder | form powder | form powder |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C |
| color yellow to brown | color white to beige | color white to beige | color white to beige |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



