Skip to Content

SML3333

Dianhydrogalactitol

≥95% (HPLC)

Synonym(s):

1,2:5,6-Dianhydrogalactitol, 1,2:5,6-Diepoxydulcitol, DAG, NSC-132313, VAL-083

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H10O4
CAS Number:
Molecular Weight:
146.14
UNSPSC Code:
12352200
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O[C@H]([C@@]1(CO1)[H])[C@H]([C@]2(CO2)[H])O

InChI

1S/C6H10O4/c7-5(3-1-9-3)6(8)4-2-10-4/h3-8H,1-2H2/t3-,4+,5+,6-

InChI key

AAFJXZWCNVJTMK-GUCUJZIJSA-N

Biochem/physiol Actions

Blood brain barrier penetrant alkylating agent that causes N7-guanine alkylation
Dianhydrogalactitol is a blood brain barrier penetrant and potent alkylating agent that causes N7-guanine alkylation and inter-strand DNA crosslinks that induces cell cycle arrest followed by apoptosis. Dianhydrogalactitol exhibits potent antitumor activity in number of clinical models including metastatic NSCLC resistant to TKI and glioblastoma multiforme.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library