Skip to Content
Merck

SML3541

KM04794

≥95% (HPLC)

Synonym(s):

3-(Trifluoromethyl)benzoic acid 2-[2-[2-[[(4-chlorophenyl)methyl]thio]phenoxy]acetyl]hydrazid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H18ClF3N2O3S
CAS Number:
Molecular Weight:
494.91
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NNC(C1=CC=CC(C(F)(F)F)=C1)=O)COC2=CC=CC=C2SCC3=CC=C(Cl)C=C3

InChI

1S/C23H18ClF3N2O3S/c24-18-10-8-15(9-11-18)14-33-20-7-2-1-6-19(20)32-13-21(30)28-29-22(31)16-4-3-5-17(12-16)23(25,26)27/h1-12H,13-14H2,(H,28,30)(H,29,31)

InChI key

CTHFEQURMFRYPT-UHFFFAOYSA-N

Biochem/physiol Actions

Cell penetrant and potent a chemical chaperone-like modulator of endoplasmic reticulum proteostasis that alleviates ER stress.
KM04794 is a cell penetrant and potent a chemical chaperone-like modulator of endoplasmic reticulum proteostasis that alleviates ER stress. It prevents protein aggregation and cell death caused by ER stressors (such as thapsigargin and dithiothreitol) and a mutant insulin. KM04794 enhances insulin production in pancreatic β cells. Apparently it binds directly to ER molecular chaperone BiP (binding immunoglobulin protein).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library