Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C29H39Cl2N3O7
CAS Number:
Molecular Weight:
612.54
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
CC1C(C(OCCOC2=CC(Cl)=C(C=C2)Cl)=O)=C(NC(C)=C1C(OC(CN3CCOCC3)CN4CCOCC4)=O)C
InChI
1S/C29H39Cl2N3O7/c1-19-26(28(35)40-15-14-39-22-4-5-24(30)25(31)16-22)20(2)32-21(3)27(19)29(36)41-23(17-33-6-10-37-11-7-33)18-34-8-12-38-13-9-34/h4-5,16,19,23,32H,6-15,17-18H2,1-3H3
InChI key
WZWJKBCMEZITJW-UHFFFAOYSA-N
Biochem/physiol Actions
Highly potent and selective platelet-activating factor (PAF) receptor antagonist
LAU-0901 is a highly potent and selective platelet-activating factor (PAF) receptor antagonist that is neuroprotective in experimental stroke, and protective in a variety of inflammation models. LAU-0901 blocks pro-inflammatory signaling. LAU-0901 demonstrated efficacy in a mouse model of glioblastoma multiforme (GBM) alone and in combination with Avastin.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.