Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C22H15F3N2O3
CAS Number:
Molecular Weight:
412.36
UNSPSC Code:
12352200
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
SMILES string
O=C1N(C(C=C(N1)C(F)(F)F)=O)C2=CC(OC3=CC=CC=C3C)=CC4=C2C=CC=C4
InChI key
LZLNIHXKWZZQLE-UHFFFAOYSA-N
Biochem/physiol Actions
BAY-771 is an inactive analog of and a negative control for the potent and selective branched chain amino acid (BCAA) transaminase (BCAT) inhibitor BAY-069 (BCAT1/2 IC50 = 6.5/10.8 μM vs 27/130 nM when using BAY-069).
Inactive analog of and negative control for the potent and selective branched chain amino acid transaminase 1/2 (BCAT1 & BCAT2) inhibitor BAY-069.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.