Skip to Content

SML3647

Ulifloxacin

≥98% (HPLC)

Synonym(s):

6-Fluoro-1-methyl-4-oxo-7-(1-piperazinyl)-1H,4H-[1,3]thiazeto[3,2-a]quinoline-3-carboxylic acid, AF 3013, NAD 394, NM 394

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H16FN3O3S
CAS Number:
Molecular Weight:
349.38
UNSPSC Code:
51111800
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1cc2c(cc1N4CCNCC4)N3C(SC3=C(C2=O)C(=O)O)C

InChI

1S/C16H16FN3O3S/c1-8-20-11-7-12(19-4-2-18-3-5-19)10(17)6-9(11)14(21)13(16(22)23)15(20)24-8/h6-8,18H,2-5H2,1H3,(H,22,23)

InChI key

SUXQDLLXIBLQHW-UHFFFAOYSA-N

Biochem/physiol Actions

Orally available, broad-spectrum fluoroquinolone antibacterial agent that potently inhibits bacterial DNA gyrase and topoisomerase IV
Ulifloxacin is an orally available, broad-spectrum fluoroquinolone antibacterial agent. Ulifloxacin is an active metabolite of Prulifloxacin. It potently inhibits bacterial DNA gyrase and topoisomerase IV.


Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Lact. - Skin Irrit. 2 - STOT RE 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library