Skip to Content

SML3683

HHV maturational protease inhibitor 43

≥97% (1H-NMR)

Synonym(s):

(1-(3-Chloro-2-oxopropyl)-4-phenyl-1H-1,2,3-triazol-5-yl)methyl benzylcarbamate, HHV inhibitor 43

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H19ClN4O3
Molecular Weight:
398.84
NACRES:
NA.77
UNSPSC Code:
51111800
Assay:
≥97% (1H-NMR)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (1H-NMR)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Covalent inhibitor of human herpesviruses (HHV) maturational protease.
HHV maturational protease inhibitor 43 is a cell penetrant, potent and selective covalent inhibitor of human herpesviruses (HHV) maturational protease that binds to non-catalytic Cysteine 161 adjacent to the protease active site. HHV maturational protease Inhibitor 43 disrupts HCMV (human cytomegalovirus) replication and infectivity. It can be used for managing human herpesviruses (HHV) infection.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library