Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C22H27FN4O
CAS Number:
Molecular Weight:
382.47
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
O=C1NC2=C(N1CCCCCN3CCN(CC3)C4=CC=C(C=C4)F)C=CC=C2
Biochem/physiol Actions
β-arrestin-biased 5-HT7-selective agonist with antagonist/inverse agonist activity against Gs signaling and in vivo antinociceptive efficacy in mouse pain models.
Serodolin is a β-arrestin-biased, 5-HT7-selective 5-hydroxytryptamine receptor agonist (5HT7/2A/1A/6 Ki = 6.2/19/454/1224 nM) that induces ERK activation through a β-arrestin-dependent signaling mechanism that requires c-SRC activation (309% of basal pErk in 5HT7-transfected HEK293 post 2-15 min 10 μM treatment) while acting as an antagonist/inverse agonist against Gs signaling (IC50 = 14 & 5 nM, respectively, against basal and 10 nM 5-CT-induced cAMP production in 5HT7-transfected HEK293). Serodolin exhibits antinociceptive efficacy in response to inflammatory, thermal, and mechanical stimulation in mice in vivo (0.1-10 mg/kg s.c.).
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.