Skip to Content

SML3719

SIAIS630121-NC

≥98% (HPLC)

Synonym(s):

(E)-N-(4-(1-(4-(4-(8-(2-(1-methyl-2,6-dioxopiperidin-3-yl)-1-oxoisoindolin-4-yl)oct-7-yn-1-yl)piperazin-1-yl)benzoyl)piperidin-4-yl)butyl)-3-(pyridin-3-yl)acrylamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C50H61N7O5
Molecular Weight:
840.06
UNSPSC Code:
51111800
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(/C=C/C1=CN=CC=C1)NCCCCC2CCN(CC2)C(C3=CC=C(C=C3)N4CCN(CC4)CCCCCCC#CC5=CC=CC6=C5CN(C6=O)C7C(N(C(CC7)=O)C)=O)=O

InChI key

ZSYJCJILRORWPM-HAVVHWLPSA-N

Biochem/physiol Actions

Negative control for NAMPT degrader SIAIS630121
SIAIS630121-NC is a negative control for NAMPT (nicotinamide phosphoribosyltransferase) degrader SIAIS630121 containing methylated CRBN ligand.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library