Skip to Content

SML3720

PHA-793887

≥98% (HPLC)

Synonym(s):

N-{6,6-Dimethyl-5-[(1-methylpiperidin-4-yl)carbonyl]-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazol-3-yl}-3-methylbutanamide, PHA 793887, PHA793887

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H31N5O2
CAS Number:
Molecular Weight:
361.48
UNSPSC Code:
12352200
NACRES:
NA.21
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N1(CCC(CC1)C(=O)N2C(c3n[nH]c(c3C2)NC(=O)CC(C)C)(C)C)C

InChI

1S/C19H31N5O2/c1-12(2)10-15(25)20-17-14-11-24(19(3,4)16(14)21-22-17)18(26)13-6-8-23(5)9-7-13/h12-13H,6-11H2,1-5H3,(H2,20,21,22,25)

InChI key

HUXYBQXJVXOMKX-UHFFFAOYSA-N

Biochem/physiol Actions

Highly potent and selective pan-CDK inhibitor with anti-cancer efficacy in cultures and in vivo.
PHA-793887 is a highly potent and selective pan-CDK inhibitor (CDK1/2//4/5/7/9 IC50 5-140 nM; IC50 >10 μM against 34 other kinases) with anti-proliferative activity against several solid tumor cell lines (IC50 <1 μM) by blocking Rb phosphorylation and the expression of S-phase cyclins, such as cyclin A. PHA-793887 exhibits in vivo anti-leukemia efficacy in murine xenograft models (20 mg/kg via daily i.v.; HL60, K562, ALL-2 or AML-PS).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library