Skip to Content

SML3738

AS2717638

≥98% (HPLC)

Synonym(s):

6,7-Dimethoxy-2-(5-methyl-1,2-benzisoxazol-3-yl)-4-(1-piperidinylcarbonyl)-1(2H )-isoquinolinone, 6,7-Dimethoxy-2-(5-methyl-1,2-benzoxazol-3-yl)-4-(piperidin-1-ylcarbonyl)isoquinolin-1(2H)-one, AS 2717638, AS-2717638

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H25N3O5
CAS Number:
Molecular Weight:
447.48
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

O=C1C2=C(C(C(N3CCCCC3)=O)=CN1C4=NOC5=C4C=C(C)C=C5)C=C(OC)C(OC)=C2

InChI

1S/C25H25N3O5/c1-15-7-8-20-18(11-15)23(26-33-20)28-14-19(24(29)27-9-5-4-6-10-27)16-12-21(31-2)22(32-3)13-17(16)25(28)30/h7-8,11-14H,4-6,9-10H2,1-3H3

InChI key

QRMYHYZQUGJBBX-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active, potent and selective lysophosphatidic acid (LPA) receptor LPAR5 (LPA5) antagonist with analgesic efficacy in vivo.

AS2717638 is an orally active, potent and selective lysophosphatidic acid (LPA) receptor LPAR5 (LPA5) antagonist (h/m/r LPA5 Ki = 9.1/7.3/16 nM) that blocks LPA-induced activation of human LPA5-expressing cells (cAMP accumulation IC50 = 38 nM), but not LPA1, LPA2 or LPA3 transfectants (Ca2+ mobilization IC50 >10 µM) and displays much reduced or little affinity toward 20 other receptors and channels. AS2717638 exhibits analgesic efficacy in mice (10 & 30 mg/kg p.o. against allodynia caused by 10 ng PGE2, 300 ng PGF2α, or 3 ng AMPA per mouse via i.t.) and rats (10 mg/kg p.o. against mechanical allodynia, thermal hyperalgesia and adjuvant-induced inflammatory pain) in vivo.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library