Skip to Content

SML3749

UCM-1306

≥98% (HPLC)

Synonym(s):

2-(Fluoromethoxy)-4′-(S-methylsulfonimidoyl)biphenyl, 2-(Fluoromethoxy)-4′-(S-methanesulfonimidoyl)-1,1′-biphenyl, UCM 1306, UCM1306

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H14FNO2S
CAS Number:
Molecular Weight:
279.33
NACRES:
NA.28
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

Orally active dopamine receptor D1-selective postive allosteric modulator (PAM) in vitro and in vivo.

UCM-1306 is an orally active dopamine receptor D1-selective postive allosteric modulator (PAM) void of D1 agonist activity. UCM-1306 potentiates dopamine (DA)-induced cAMP in the SK-N-MC neuroblastoma cell line with endogenous D1R (EC50/Emax = 60 nM/45% with [DA] = EC70; Emax = 55% with [DA] = 10 μM), but not D5R, by modulating DA affinity for the receptor, while displaying low binding competition against orthosteric ligands. UCM-1306 augments cocaine (20 mg/kg s.c.)-induced locomotion (1 mg/kg i.p.) and enhances L-DOPA (100-400 mg/kg i.p.) recovery of decreased locomotor activity in reserpinized (2.5 mg/kg s.c.) mice in vivo (30 mg/kg p.o.).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library