Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C55H76N10O12S
Molecular Weight:
1101.32
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
powder
Quality Segment
assay
≥95% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
SMILES string
O=C(C1=NC=NC(NC2CN(C(COCCOCCOCCOCCOCC(N[C@H](C(N3[C@@H](C(N[C@H](C4=CC=C(C5=C(C)N=CS5)C=C4)C)=O)C[C@H](O)C3)=O)C(C)(C)C)=O)=O)C2)=C1)NC[C@H](O)CN6CC7=C(C=CC=C7)CC6
InChI
1S/C55H76N10O12S/c1-36(38-10-12-40(13-11-38)50-37(2)59-35-78-50)60-53(71)46-24-43(66)31-65(46)54(72)51(55(3,4)5)62-48(68)32-76-22-20-74-18-16-73-17-19-75-21-23-77-33-49(69)64-28-42(29-64)61-47-25-45(57-34-58-47)52(70)56-26-44(67)30-63-15-14-39-8-6-7-9-41(
InChI key
XUJMNOQMXWVXQE-KPPAQSJOSA-N
Biochem/physiol Actions
Inactive control for PRMT5 degrader MS4322.
MS4370 is an inactive control for PRMT5 degrader MS4322. It comprises of PRMT5 inhibitor EPZ015666 is linked to a von Hippel-Lindau (VHL) E3 ligase ligand (R,S,S)-AHPC-Me (diastereoisomer of VHL-2) that acts as a potent, cell-permeable PRMT5 inhibitor (IC50 = 12 nM). MS4730 does neither degrade PRMT5 nor reduce PRMT5 protein levels at 5 µM, and thus serves as a suitable control for MS4322.
MS4370 is an inactive control for PRMT5 degrader MS4322. It comprises of PRMT5 inhibitor EPZ015666 is linked to a von Hippel-Lindau (VHL) E3 ligase ligand (R,S,S)-AHPC-Me (diastereoisomer of VHL-2) that acts as a potent, cell-permeable PRMT5 inhibitor (IC50 = 12 nM). MS4730 does neither degrade PRMT5 nor reduce PRMT5 protein levels at 5 µM, and thus serves as a suitable control for MS4322.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.