Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C34H27NO5S2
CAS Number:
Molecular Weight:
593.71
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
VDAC2 ligand that enhances VDAC2:BAK interaction and selectively blocks murine BAK-mediated apoptosis.
WEHI-9625 is a VDAC2 ligand that enhances VDAC2:BAK interaction and selectively blocks murine BAK-mediated apoptosis in early stages. WEHI-9625 preserves cellular function and long-term clonogenic potential before apoptosis induced mitochondrial damage. WEHI-9625 is inactive against both human BAK (hBAK) and BAX.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.