Skip to Content

SML3918

MKA031

≥98% (HPLC)

Synonym(s):

1-(4-(Phenylcarbamoyl)phenyl)-N-(thiophen-2-ylmethyl)-1H-1,2,3-triazole-4-carboxamide, MKA 031, MKA-031

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H17N5O2S
CAS Number:
Molecular Weight:
403.46
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=C(C1=CN(C2=CC=C(C(NC3=CC=CC=C3)=O)C=C2)N=N1)NCC4=CC=CS4

Biochem/physiol Actions

Allosteric site-targeting macrophage migration inhibitory factor (MIF) inhibitor that protects HeLa cells against MNNG-induced parthanatos.
MKA031 is an allosteric site-targeting macrophage migration inhibitory factor inhibitor (MIF tautomerase activity IC50 = 1.7 μM) that prevents MNNG-induced MIF nuclear translocation (3h 100 μM MKA031 preincubation prior to 15-min 50 μM MNNG treatment; HeLa cells) by interfering with MIF/AIF interaction (IC50 = 12.3 μM against 4 μM AIF in wells precoated with 100 μL of a 250 nM MIF solution). Unlike ZP143, MKA031 protects HeLa cells against MNNG-induced parthanatos (EC50 = 7.7 μM 24h post 15-min 50 μM MNNG treatment).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library