Skip to Content

SML3934

JA397

≥98% (HPLC)

Synonym(s):

tert-butyl 3-((2-((4-((tert-butoxycarbonyl)amino)benzyl)amino)pyrimidin-4-yl)amino)-1H-pyrazole-5-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H31N7O4
Molecular Weight:
481.55
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OC(C)(C)C)C1=CC(NC2=NC(NCC3=CC=C(NC(OC(C)(C)C)=O)C=C3)=NC=C2)=NN1

Biochem/physiol Actions

Cell penetrant, potent and selective inhibitor CDK14-18 (TAIRE family).
JA397 is a cell penetrant, potent and selective inhibitor for the TAIRE family of the cyclin-dependent kinases (CDK14-18).


hcodes

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library