Skip to Content

SML3938

H55

≥98% (HPLC)

Synonym(s):

(Z)-4-(4-(4-bromophenyl)-2-(phenylimino)thiazol-3(2H)-yl)-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one, Erg6 inhibitor H55

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H21BrN4OS
CAS Number:
Molecular Weight:
517.44
NACRES:
NA.77
UNSPSC Code:
51111800
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

Biochem/physiol Actions

Potent and selective allosteric inhibitor of the fungal-specific enzyme Erg6 that induces accumulation of zymosterol and farnesol in pathogenic fungus C. albicans.


H55 is a cell penetrant, potent and selective allosteric inhibitor of the fungal-specific enzyme Erg6 (C-24 sterol methyltransferase) that induces accumulation of zymosterol and farnesol in pathogenic fungus Candida albicans. H55 potently inhibits C. albicans hyphal formation. Treatment of mice infected with C. albicans with H55 alleviates the severity of the infection and improves the survival.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library