Skip to Content

SML3939

Org 34167

≥98% (HPLC)

Synonym(s):

(αS)-2-(1,2-Benzisoxazol-3-yl)-α-2-propen-1-ylbenzenemethanamine, (1S)-1-(2-(1,2-Benzoxazol-3-yl)phenyl)but-3-en-1-amine, (S)- 2-(1,2-Benzisoxazol-3-yl)-α-2-propenyl-benzenemethanamine, ORG-34167

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H16N2OHCl
CAS Number:
Molecular Weight:
300.78
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

C=CC[C@H](N)C(C=CC=C1)=C1C2=NOC3=CC=CC=C32.Cl

Biochem/physiol Actions

Brain penetrant, potent and selective modulator of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels that exhibit antidepressant activity.
Org 34167 is a brain penetrant, potent and selective modulator of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels that exhibit antidepressant activity in human and rat models. Apparently, Org 34167 inhibits HCN1 channel activity by modulating hyperpolarizing shift in activation voltage dependence and a slowing of activation kinetics.

Disclaimer

Hygroscopic


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library