Skip to Content

SML3966

Arborinine

≥98% (HPLC)

Synonym(s):

1-Hydroxy-2,3-dimethoxy-10-methyl-9(10H)-acridinone, 1-Hydroxy-2,3-dimethoxy-10-methylacridan-9-one, 1-Hydroxy-2,3-dimethoxy-10-methylacridone, Arborinin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H15NO4
CAS Number:
Molecular Weight:
285.29
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C1C2=C(C=C(C(OC)=C2O)OC)N(C)C3=C1C=CC=C3

Biochem/physiol Actions

Alkaloid isolated from leaves of Glycosmis bilocularis that exhibits strong anti-cancer, anti-bacterial, and anti-viral activities.
Arborinine is a quinoline alkaloid isolated from leaves of Glycosmis bilocularis that exhibits strong anti-cancer, anti-bacterial, and anti-viral activities. Arborinine is a broad-spectrum antiproliferative agent, which potently inhibits proliferation of numerous cancer cell lines including NCI-N87 (IC50 = 5.67 μM), BGC-823 (IC50 = 7.26 μM), MGC803 (IC50 = 4.75 μM) and more. It potently inhibits SKOV3 ovarian carcinoma cell proliferation and tumor growth. It also reduces migration and invasion of SKOV3 cancer cells. Arborinine reduces expression of histone demethylase LSD1, which leads to accumulation of H3K4m1.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library