Skip to Content

SML3986

JP3001

≥98% (HPLC)

Synonym(s):

JP 3001, JP-3001, JP 3000 inactive control, JP-3000 inactive control, JP3000 inactive control, JP 3000 negative control, JP-3000 negative control, JP3000 negative control, trans-2-(5-(3,5-bis(Trifluoromethyl)phenyl)-4-phenyloxazol-2-yl)cyclopropyl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H15F6NO2
CAS Number:
Molecular Weight:
427.34
NACRES:
NA.28
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC[C@H](C1)[C@@H]1C(O2)=NC(C3=CC=CC=C3)=C2C4=CC(C(F)(F)F)=CC(C(F)(F)F)=C4

Biochem/physiol Actions

Inactive analog of and the recommended negative control for the potent and selective retinoid X receptors (RXRs) agonist JP3000.
JP3001 is an inactive analog of and the recommended negative control (RXRα/β/γ EC50 >10 µM) for the potent and selective retinoid X receptors (RXRs) agonist JP3000.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library