Skip to Content

SML4004

pan-Orai inhibitor, 5D

≥98% (HPLC)

Synonym(s):

N-[2,2,2-Trichloro-1-(2-naphthalenylamino)ethyl]-2-furancarboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H13Cl3N2O2
CAS Number:
Molecular Weight:
383.66
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=CO1)NC(NC2=CC=C3C(C=CC=C3)=C2)C(Cl)(Cl)Cl

Biochem/physiol Actions

Potent blocker of all Orai channels indiscriminately with an IC50 value of 195 nM in blocking SOCE in primary T cells.

Pan-Orai inhibitor 5D is a potent blocker of all Orai channels indiscriminately with an IC50 value of 195 nM in blocking SOCE in primary T cells. pan-Orai inhibitor 5D potently inhibits effector T-cell function and IL-2 production.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library