Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C25H33F2IN6
Molecular Weight:
582.47
Assay:
≥90% (HPLC)
Form:
powder
Storage condition:
desiccated, protect from light
Quality Segment
assay
≥90% (HPLC)
form
powder
storage condition
desiccated, protect from light
color
faint red to black
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
Biochem/physiol Actions
MAO-N3, a cell-permeant HIDE (High-Density Environmentally-sensitive) probe, is tailored for precise targeting of the inner mitochondrial membrane, and when combined with SiR-DBCO, it forms the MAO-SiR complex, the anticipated product of strain-promoted azide–alkyne click chemistry (SPAAC). MAO-SiR exhibits exceptional photostability, significantly reducing photobleaching issues in extended imaging sessions. Its non-toxic nature ensures safe application across various cell types, including delicate primary cardiomyocytes. Furthermore, MAO-N3′s compatibility with sophisticated imaging techniques such as confocal, SIM, SMLM, and STED microscopy, facilitates detailed visualization of complex mitochondrial structures, like cristae, with superior spatial and temporal resolution. Note: In time-lapse imaging, cells treated with 200 nM MAO-N3 and 100 nM SiR-DBCO, collectively known as MAO-SiR, exhibit minimal signal degradation and a rise in cell density, indicative of robust mitochondrial function and cell health. This outcome is in stark contrast to the results observed with dyes like MitoTracker Deep Red, which typically show a substantial decline in signal intensity within just a few hours.
Disclaimer
Hygroscopic Store with dessicant
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Skin Irrit. 2
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
