Skip to Content

SML4108

Anethole Trithione

≥98% (HPLC)

Synonym(s):

3-(p-Methoxyphenyl)trithione, 5-(4-Methoxyphenyl)-3H-1,2-dithiole-3-thione, Anethole dithiolthione, OP2113

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H8OS3
CAS Number:
Molecular Weight:
240.36
MDL number:
NACRES:
NA.28
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

, faint orange to red

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

S=C1SSC(C2=CC=C(C=C2)OC)=C1

Biochem/physiol Actions

Inhibitor of mitochondrial ROS production; medication used in the treatment of dry mouth.

Anethole Trithione (OP2113) is an old medication used in the treatment of dry mouth that increases salivary secretion and blocks carcinogenesis. It blocks mitochondrial ROS production. Anethole Trithione is a specific inhibitor of ROS production at the IQ site of complex I of the mitochondrial respiratory chain. It reduces pulmonary arterial pressure, remodeling, and hyperreactivity in CH-PH rats.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library