Skip to Content

SML4117

Phyllodulcin

≥95% (HPLC)

Synonym(s):

(3R)-3,4-Dihydro-8-hydroxy-3-(3-hydroxy-4-methoxyphenyl)-1H-2-benzopyran-1-one, (3R)-8-Hydroxy-3-(3-hydroxy-4-methoxyphenyl)-3,4-dihydro-1H-isochromen-1-one, 3R-Phyllodulcin, d-Phyllodulcin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H14O5
CAS Number:
Molecular Weight:
286.28
MDL number:
NACRES:
NA.21
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C1C2=C(O)C=CC=C2C[C@H](C3=CC(O)=C(C=C3)OC)O1

Biochem/physiol Actions

Orally bioavailable and blood brain barrier permeable natural sweetener that mediates anti-diabetic, anti-inflammatory and anti-fungal activities.
Phyllodulcin is an orally bioavailable and blood brain barrier permeable natural sweetener isolated from Hydrangea macrophylla var. thunbergii that mediates anti-diabetic, anti-inflammatory and anti-fungal activities. Phyllodulcin efficiently reduces Aβ aggregation in the hippocampus and enhances memory-related processes. It minimizes neuroinflammation in the hippocampus and inhibits microglia and astrocytes activation.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library