Skip to Content

SML4127

BRD0539

≥98% (HPLC)

Synonym(s):

((3aR,4R,9bR)-8-(2-fluorophenyl)-1-tosyl-2,3,3a,4,5,9b-hexahydro-1H-pyrrolo[3,2-c]quinolin-4-yl)methanol, (3aR,4R,9bR)-8-(2-Fluorophenyl)-2,3,3a,4,5,9b-hexahydro-1-[(4-methylphenyl)sulfonyl]-1H-pyrrolo[3,2-c]quinoline-4-methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H25FN2O3S
CAS Number:
Molecular Weight:
452.54
MDL number:
NACRES:
NA.28
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

OC[C@@H]1NC2=CC=C(C3=C(F)C=CC=C3)C=C2[C@H]4[C@@H]1CCN4S(C5=CC=C(C)C=C5)(=O)=O

Biochem/physiol Actions

Cell penetrant and potent anti-CRISPR agent that inhibits SpCas9-DNA interaction.


BRD0539 is a cell penetrant and potent anti-CRISPR agent. BRD0539 is a reversible inhibitor of Streptococcus pyogenes Cas9 (SpCas9) that disrupt the SpCas9-DNA interaction. It potently inhibits SpCas9/PAM binding leading to a decrease in nuclease activity. BRD0539 is stable under physiological conditions.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library