Skip to Content

SML4128

K-975

≥95% (HPLC)

Synonym(s):

K 975, K975, N-[3-(4-Chlorophenoxy)-4-methylphenyl]-2-propenamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H14ClNO2
CAS Number:
Molecular Weight:
287.74
MDL number:
NACRES:
NA.28
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(C)C(OC2=CC=C(C=C2)Cl)=C1)C=C

Biochem/physiol Actions

Orally active, potent, irreversible pan-TEAD inhibitor that disrupts YAP1/TAZ-TEAD association and inhibits malignant pleural mesothelioma proliferation in vitro and in vivo.


K-975 is an orally active, potent, irreversible pan-TEAD inhibitor that disrupts YAP1/TAZ-TEAD interaction via covalent modification of TEAD palmitate-binding pocket (PBP) cysteine. K-975 suppresses NCI-H661 cellular TEAD transcriptional activity (IC50 ~100 nM by reporter assay) and selectively inhibits the proliferations of neurofibromin 2 (NF2)-deficient malignant pleural mesothelioma (MPM) cultures (IC50 ≤175 nM) over NF2-expressing MPM. K-975 suppressed tumor growth in MPM xenograft models in mice in vivo (35-300 mg/kg b.i.d. p.o. for 4 wks).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library