Skip to Content

SML4134

AJ2-30

≥98% (HPLC)

Synonym(s):

9-Ethyl-N-(1-methyl-1H-benzimidazol-2-yl)-9H-carbazole-3-methanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H22N4
CAS Number:
Molecular Weight:
354.45
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCN1C2=C(C=CC=C2)C3=CC(CNC4=NC5=C(N4C)C=CC=C5)=CC=C31

Biochem/physiol Actions

Cell penetrant, potent and selective first-in-class functional inhibitors of SLC15A4.

AJ2-30 is a cell penetrant, potent and selective first-in-class functional inhibitors of SLC15A4 (IC50 = 1.8 μM (IFN-α suppression) and IC50 = 2.6 μM (MDP transport inhibition)) that disrupt all functions of SLC15A4. AJ2-30 potently suppressed IFN-α production in human pDCs after stimulation with TLR7/8 and TLR9 agonists. It significantly suppressed production of IFN-γ, IL-6 and IL-10 in lupus patient-derived PBMCs, and significantly reduced IFN-α, IFN-β and IFN-γ production in mice.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library