Skip to Content

SML4151

BI-2865

≥98% (HPLC), powder

Synonym(s):

(4S)-2-Amino-4,5,6,7-tetrahydro-4-methyl-4-[3-[4-[(1S)-1-[(2S)-1-methyl-2-pyrrolidinyl]ethoxy]-2-pyrimidinyl]-1,2,4-oxadiazol-5-yl]- benzo[b]thiophene-3-carbonitrile, (S)-2-amino-4-methyl-4-(3-(4-((S)-1-((S)-1-methylpyrrolidin-2-yl)ethoxy)pyrimidin-2-yl)-1,2,4-oxadiazol-5-yl)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H27N7O2S
CAS Number:
Molecular Weight:
465.57
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

NC(S1)=C(C#N)C2=C1CCC[C@]2(C)C3=NC(C4=NC(O[C@@H](C)[C@]5([H])CCCN5C)=CC=N4)=NO3

Application

BI-2865 may be used for studying the Kirsten rat sarcoma virus (KRAS)signaling pathways and their role in cancer. Its selective inhibition of KRAS isused to probe the biological functions of KRAS, its mutants, and downstreamsignaling pathways to gain a deeper understanding of KRAS-related oncogenesis.

Biochem/physiol Actions

BI-2865 is a cell penetrant, selective and potent pan-KRAS inhibitor, which binds with high affinity to the inactive state of KRAS and KRAS mutants. BI-2865 blocks nucleotide exchange to prevent the activation of wild-type KRAS and a broad range of KRAS mutants. It inhibits downstream signaling and growth in cancer cells harboring mutant KRAS. It does not bind to NRAS and HRAS.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library