Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C12H14O5
CAS Number:
Molecular Weight:
238.24
MDL number:
NACRES:
NA.21
Assay:
≥95% (HPLC)
Form:
powder
Quality Segment
assay
≥95% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
SMILES string
OC(CC(CC1=CC=CC(OC)=C1)C(O)=O)=O
Application
SLC13A3i may be used in studies for its role in inhibiting the uptake of itaconate in tumor cells, which may help understand tumor immune evasion mechanisms and improve cancer treatment strategies. It may also be used to explore metabolic pathways related to integrin functions and their impact on cellular processes, particularly in cancer research.
Biochem/physiol Actions
A cell-permeable solute carrier family 13-member 3 (SLC13A3) inhibitor (SLC13A3i) targets the SLC13A3 transporter to block the oncometabolite itaconate uptake, thereby reducing tumor ferroptosis resistance and enhancing immune responses in SLC13A3+ tumors. By disrupting the NRF2-SLC7A11 pathway, SLC13A3i sensitizes tumor cells to ferroptosis and increases CD8+ T cell infiltration in the tumor microenvironment. Preclinical studies show robust efficacy of SLC13A3i in both in vitro (1-5 mM) and in vivo (10-20 mg/kg, i.p.) models, especially when combined with anti-PD-L1 immunotherapy, resulting in slowed tumor progression and improved immune cell engagement. With a favorable safety profile and metabolic stability, SLC13A3i emerges as a promising immunometabolic therapeutic candidate for treating resistant cancers.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
