Skip to Content

SML4172

Iodoacetamide Alkyne-C3

≥95% (HPLC), powder, acysteine-reactive ABPP benchmark probe

Synonym(s):

IPM, IA-Alkyne-C3, IAM-Alkyne-C3, IAA-C3, N-Propynyliodoacetamide, 2-Iodo-N-(prop-2-yn-1-yl)acetamide, 2-Iodo-N-2-propyn-1-ylacetamide, N-Propargyl-2-iodoacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H6INO
CAS Number:
Molecular Weight:
223.01
MDL number:
NACRES:
NA.21
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(CI)NCC#C

Application

Iodoacetamide Alkyne-C3 is suitable for use in isotopic tagging strategies for quantitative cysteine-reactivity profiling in various biological samples.

Biochem/physiol Actions

Iodoacetamide alkyne-C3 (IPM, IAA-C3, IA-alkyne-C3, IAM-alkyne-C3) is a bifunctional, thiol-reactive probe engineered for activity-based protein profiling (ABPP) of reactive cysteine residues in live cells, tissue homogenates, and tissues. This probe combines an iodoacetamide group, functioning as a covalent warhead for selective cysteine labeling, with a C3 alkyne moiety that acts as a click-chemistry handle. This alkyne allows for the subsequent conjugation of fluorophores or biotin-linked azido reporters, facilitating either the visualization or enrichment of labeled proteins. The IAA-C3 probe, the gold standard in the cysteinomics field, facilitates site-specific analysis of cysteine accessibility, reactivity, redox modification status, and covalent engagement in biological samples by allowing researchers to treat cell homogenates with varying concentrations of IA-alkyne C3, identifying highly reactive cysteines-such as those in enzyme active sites or redox-sensitive regions-that remain reactive despite dilution effects.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library