Skip to Content

SML4190

Ropidoxuridine

≥98% (HPLC), powder, radiosensitizer

Synonym(s):

5-Iodo-2-pyrimidinone-2′-deoxyribose, 1-(2-Deoxy-β-D-erythro-pentofuranosyl)-5-iodo-2(1H)-pyrimidinone, 1-(2-Deoxy-β-D-ribofuranosyl)-5-iodo-2-pyrimidinone, IPdR

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H11IN2O4
CAS Number:
Molecular Weight:
338.10
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1N(C=C(C=N1)I)[C@@H]2O[C@@H]([C@H](C2)O)CO

General description

Ropidoxuridine, a thymidine analog, is an orally available pro-drug of 5-iodo-2′-deoxyuridine (IUdR).

Application

Ropidoxuridine is suitable for use in radiosensitization studies in vivo using cancer models and cell lines.

Biochem/physiol Actions

Ropidoxuridine, a radiation sensitizer, after oral administration it is metabolized by an aldehyde oxidase in the liver to the active drug. Ropidoxuridine sensitizes cancer cells to radiation.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library