Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C16H28O2
CAS Number:
Molecular Weight:
252.39
MDL number:
NACRES:
NA.21
Assay:
≥95% (HPLC)
Form:
powder
Quality Segment
assay
≥95% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
SMILES string
O=C(CCCCCCCCCCCCCC#C)O
General description
Alkynyl palmitic acid (Alk-C16) is a modified palmitic acid consisting of a palmitic acid backbone with ω-terminal alkyne. This structure allows it to participate in bio-orthogonal reactions, particularly in copper-catalyzed click chemistry.
Application
Alk-C16 is suitable for use as a palmitoylation probe in:
- fluorescence imaging to study palmitoylation dynamics during the cell cycle
- characterizing the mechanisms of pharmacological inhibitors of fatty acyltransferases
- biochemical and imaging detection of palmitoylated proteins in cells
Biochem/physiol Actions
Bio-orthogonal probe used for metabolic labeling and fluorescence imaging of palmitoylated proteins.
Alkynyl Palmitic Acid (Alk-C16) is a bio-orthogonal probe designed for metabolic labeling of native palmitoylated proteins both in vitro and in vivo. It enables precise and chemoselective labeling via Cu(I)-catalyzed click chemistry, which involves the reaction of Alk-C16 with suitably tagged azides. This reaction allows for the efficient conjugation of fluorescent or biotinylated azides, facilitating high-resolution fluorescence imaging to reveal the subcellular localization of palmitoylated proteins and their association with cellular membranes. Alk-C16 specifically labels cysteine residues of palmitoylated proteins, making it particularly useful for studying dynamic palmitoylation and protein trafficking. Tools like Alk-C16 play a critical role in the identification and enrichment of lipidated proteins, advancing our ability to isolate and study palmitoylated proteins in complex biological systems.
Alkynyl Palmitic Acid (Alk-C16) is a bio-orthogonal probe designed for metabolic labeling of native palmitoylated proteins both in vitro and in vivo. It enables precise and chemoselective labeling via Cu(I)-catalyzed click chemistry, which involves the reaction of Alk-C16 with suitably tagged azides. This reaction allows for the efficient conjugation of fluorescent or biotinylated azides, facilitating high-resolution fluorescence imaging to reveal the subcellular localization of palmitoylated proteins and their association with cellular membranes. Alk-C16 specifically labels cysteine residues of palmitoylated proteins, making it particularly useful for studying dynamic palmitoylation and protein trafficking. Tools like Alk-C16 play a critical role in the identification and enrichment of lipidated proteins, advancing our ability to isolate and study palmitoylated proteins in complex biological systems.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
No data available
flash_point_c
No data available
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.