Skip to Content

SML4200

Empagliflozin

≥98% (HPLC)

Synonym(s):

1-Chloro-4-(β-D-glucopyranos-1-yl)-2-[4-((S)-tetrahydrofuran-3-yl-oxy)-benzyl]-benzene, (1S)-1,5-Anhydro-1-C-[4-chloro-3-[[4-[[(3S)-tetrahydro-3-furanyl]oxy]phenyl]methyl]phenyl]-D-glucitol, (2S,3R,4R,5S,6R)-2-(4-Chloro-3-(4-((S)-tetrahydrofuran-3-yloxy]benzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, BI 10773, BI-10773, BI10773

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H27ClO7
CAS Number:
Molecular Weight:
450.91
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

General description

Empagliflozin is a C-glucoside with a carbon-carbon bond between the glucose and aglycone, resisting the breakdown in the gastrointestinal tract.

Application

Empagliflozin may be used to study its effects on diabetes mellitus in animal models.

Biochem/physiol Actions

Empagliflozin (BI 10773) is an orally active, potent and highly selective sodium glucose co-transporter-2 inhibitor (hSGLT-2 IC50 = 3.1 nM; hSGLT-1/4/5/6 IC50 = 8.3/11/1.1/2.0 µM) that targets SGLT-2 with high affinity in a glucose-competitive manner (hSGLT-2 Kd = 57/194 nM in the absence/presence of 20 mM glucose). Empagliflozin improves glycaemic control in type 2 (1-3 mg/kg via daily p.o. to Zucker diabetic fatty (ZDF) rats) and type 1 (10 mg/kg b.i.d. p.o. to streptozotocin-induced diabetic (STD) rats) diabetes mellitus models.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library