Skip to Content

SML4204

Yaddle1

≥98% (HPLC), powder, Piezo1 Agonist

Synonym(s):

2-{[2-Chloro-6-(trifluoromethyl)benzyl]thio}-5-(pyrazin-2-yl)-1,3,4-oxadiazole, Pyrazine, 2-[5-[[[2-chloro-6-(trifluoromethyl)phenyl]methyl]thio]-1,3,4-oxadiazol-2-yl]- (ACI)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H8ClF3N4OS
CAS Number:
Molecular Weight:
372.75
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

General description

Yaddle1 is designed to interact specifically with the Piezo1 channel. It features a trifluoromethyl group, which is crucial for its agonistic activity.

Application

Yaddle1 may be used in immunology as a vaccine adjuvant, targeting optimal T cell activation, enhancing vaccine efficacy. It may also be used to study the role of piezo-type mechanosensitive ion channel component 1 in inflammatory bowel disease.

Biochem/physiol Actions

Yaddle1 is a mechanosensitive ion channel Piezo1 agonist with higher potency (Ca2+ influx EC50/Emax = 0.40 μM/2.6-fold using Piezo1 mCherry-expressing HEK293A) and greatly improved aqueous solubility (26.7 μM at pH 7.4) than Yoda1 (EC50/Emax = 0.50 μM/2.4-fold; solubility = 1.2 μM at pH 7.4). Yaddle1 binds to Piezo1 channel, stabilizing the open conformation of the Piezo1 channel and lowering the mechanical activation threshold.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library