Skip to Content

SML4239

Phenoxodiol

≥98% (HPLC), powder, tNOX (ENOX2) inhibitor

Synonym(s):

3-(4-Hydroxyphenyl)-2H-1-benzopyran-7-ol; 3-(4-Hydroxyphenyl)-2H-benzopyran-7-ol, Haginin E, Idronoxil, Dehydroequol, NV 06

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H12O3
CAS Number:
Molecular Weight:
240.25
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to pink

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Application

Phenoxodiol may be used to study immunomodulatory effects on human peripheral blood mononuclear cells in in vitro and in vivo assays.

Biochem/physiol Actions

Phenoxodiol, an isoflavene, is a broad-spectrum anti-tumor agent that causes apoptosis via both intrinsic and extrinsic pathways. It is a pan-cancer drug that targets plasma membrane electron transport (PMET). It also acts as a potent sensitizer of most standard chemotoxic agents. Phenoxodiol inhibits tumor-specific NADH oxidase tNOX (ENOX2) and DNA topoisomerase II. It blocks phosphorylation and activation of several different pro-survival signaling proteins including sphingosine kinase and Akt. It is also known to inhibit tyrosine kinases and X-linked inhibitor of apoptosis.


Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

No data available

Flash Point (°C)

No data available



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library