Skip to Content

SML4241

GSK269962A

≥98% (HPLC)

Synonym(s):

GSK-269962, N-[3-[[2-(4-Aminofurazan-3-yl)-1-ethyl-1H-imidazo[4,5-c]pyridin-6-yl]oxy]phenyl]-4-[[2-(4-morpholinyl)ethyl]oxy]benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H30N8O5
CAS Number:
Molecular Weight:
570.60
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Orally available, potent and selective ROCK1/2 inhibitor that impacting vasodilation and AML cell viability; selective probe for Rho-kinase signaling pathways.
GSK269962A (GSK269962) is a potent (ROCK1 IC50 1.6 nM; ROCK2 IC50 4-6 nM), selective, orally bioavailable inhibitor of ROCK kinases. Offering greater selectivity than Y-27632 and Fasudil, it weakly inhibits MSK1 (IC50 49 nM) and RSK1 (IC50 132 nM). As an ATP-competitive inhibitor, it blocks ROCK-mediated phosphorylation of downstream targets (e.g., MLC, MYPT1), causing significant functional outcomes including smooth muscle relaxation (vasorelaxation IC50 35 nM), actin stress fiber disruption, G2 cell cycle arrest, and apoptosis, notably in AML models (proliferation IC50 0.61-1337 nM). In vitro, it inhibits AngII-induced stress fibers and inflammatory cytokine production. With demonstrated in vivo efficacy lowering blood pressure in hypertensive rats and reducing leukemia burden in AML xenografts, GSK269962A serves as a valuable tool for studying ROCK signaling in cytoskeleton regulation, smooth muscle contraction, cell migration, proliferation, and survival.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library