Skip to Content

SML4254

AT7519

≥98% (HPLC), powder, CDK inhibitor

Synonym(s):

AT 7519, AT-7519, AT7519, N-(4-Piperidinyl)-4-(2,6-dichlorobenzoylamino)-1H-pyrazole-3-carboxamide, 4-(2,6-Dichlorobenzoylamino)-1H-pyrazole-3-carboxylic acid N-(piperidin-4-yl)amide, 4-[(2,6-Dichlorobenzoyl)amino]-N-4-piperidinyl-1H-pyrazole-3-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H17Cl2N5O2
CAS Number:
Molecular Weight:
382.24
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Application

AT7519 may be used to study its effects on cell viability and proliferation, cell cycle, apoptosis and pyroptosis in human glioblastoma cells.

Biochem/physiol Actions

AT7519 is a potent and selective cyclin dependent kinase (CDK) inhibitor (CDK5-p35/CDK2-CycA/CDK4-CycD1/CDK1-CycB IC50 = 18/44/67/190 nM; IC50 >10 μM against Aurora A, IR, MEK1, PDK1, c-abl) with antiproliferative activity in cancer cultures (IC50 = 82 nM/HCT116, 350 nM/A2780) and antitumor efficacy in mice in vivo (86% inhibition of A2780 xenograft tumor growth post 8-day treatment at 7.5 mg/kg b.i.d. i.p., 87% inhibition of HCT116 xenograft tumor growth post 10-day treatment at 9.1 mg/kg b.i.d. i.p.).


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library