Skip to Content

SML4255

JNJ-10397049

≥97% (HPLC), powder, OX2R antagonist

Synonym(s):

1-(2,4-Dibromophenyl)-3-[(4S,5S)-2,2-dimethyl-4-phenyl-1,3-dioxan-5-yl]urea, N-(2,4-Dibromophenyl)-N′-[(4S,5S)-2,2-dimethyl-4-phenyl-1,3-dioxan-5-yl]-urea, JNJ 10397049, JNJ10397049

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H20Br2N2O3
CAS Number:
Molecular Weight:
484.18
MDL number:
NACRES:
NA.21
Assay:
≥97% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Application

JNJ-10397049 may be used in proliferation assay to study its role in growth and differentiation of neural progenitor cells (NPCs).

Biochem/physiol Actions

JNJ-10397049 is a potent and subtype-selective orexin receptor 2 (OX2R) antagonist (pKi = 8.3/OX2R vs. 5.5/OX1R) with little affinity toward a panel of 50 receptors, ion channels and transporters. Blockade of OX2R by JNJ-10397049 antagonizes against orexin B-induced cellular calcium response (pKB = 7.9) and attenuates ethanol self-administration, place preference, and reinstatement in rats in vivo (1, 3, and 10 mg/kg, sc).


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library