Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C6H7N3O3
CAS Number:
Molecular Weight:
169.14
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Quality Segment
assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
Application
Biochem/physiol Actions
NSC232003 is a Ubiquitin-likewith PHD and RING Finger domains 1 (UHRF1) E3 ubiquitin ligase inhibitor thatdownregulates overall genomic DNA cytosine methylation (5 mC IC50 = 15 µM post72h treatment; U251 glioma cells) by disrupting UHRF1 interaction with DNAmethyltransferase DNMT1.
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
