Skip to Content

SML4269

740 Y-P trifluoroacetate

≥95% (HPLC)

Synonym(s):

740 YP trifluoroacetate, 740-YP trifluoroacetate, 740Y-P trifluoroacetate, 740YP trifluoroacetate, 740YPDGFR trifluoroacetate, PDGFR 740Y-P trifluoroacetate, RQIKIWFQNRRMKWKK-SDGGY(P)MDMS trifluoroacetate, RQIKIWFQNRRMKWKK-SDGGpYMDMS trifluoroacetate, Arg-Gln-Ile-Lys-Ile-Try-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-Ser-Asp-Gly-Gly-Tyr(P)-Met-Asp-Met-Ser trifluoroacetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C141H222N43O39PS3 · xC2HF3O2
CAS Number:
Molecular Weight:
3270.70 (free base basis)
Assay:
≥95% (HPLC)
Form:
(Powder or Lyophilized powder or film)
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

General description

Cell-permeable phosphatidylinositol 3-kinase (PI 3-kinase, PI3K) activator.
740 Y-P is a cell-permeable phosphatidylinositol 3-kinase (PI 3-kinase, PI3K) activator composed of the Drosophila antennapedia-derived membrane-permeant 16-mer sequence RQIKIWFQNRRMKWKK followed by a phosphotyrosine-containing, PDGFR-derived sequence SDGGY(P)MDMS that targets SH2 domains of PI3K p85 regulatory subunit. When applied to serum-starved C2 cells at 50 µg/mL, 740 Y-P is more effective than 10% FCS, 5 ng/mL bFGF or 10 ng/mL EGF in stimulating cell proliferation (% of cells in S-phase post 48h stimulation = 48.3, 42.5, 35.7 and 32.8, respectively). 740 Y-P-induced mitogenic response is blocked by wortmannin or rapamycin, but not MEK inhibitor PD98058.

Disclaimer

Hygroscopic Store with desiccant


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library