Skip to Content

SML4272

Fen1-IN-1

≥98% (HPLC), powder

Synonym(s):

1-[(2,3-Dihydro-1,4-benzodioxin-2-yl)methyl]-3-hydroxythieno[3,2-d]pyrimidine-2,4(1H,3H)-dione, 1-((2,3-Dihydrobenzo[b][1,4]dioxin-2-yl)methyl)-3-hydroxythieno[3,2-d]pyrimidine-2,4(1H,3H)-dione, Fen1 Inhibitor, AZ-13623940, AZ 13623940, LNT 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H12N2O5S
CAS Number:
Molecular Weight:
332.33
NACRES:
NA.21
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to gray

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

General description

Cell-permeable, selective and potent inhibitor of the FEN-1 and a chemopotentiator.
Fen1-IN-1 is a cell-permeable N-hydroxyimide compound that potently and selectively inhibits FEN-1 (Flap endonuclease 1) (IC50 = 11 nM), a divalent metal-dependent nuclear enzyme with the ability to cleave the 5′ overhang of branched dsDNA. FEN1-IN-1 minimally affects the activity of a related endonuclease xeroderma pigmentosum G (XPG; IC50 = 292 nM). Dose-dependently potentiates methyl methanesulfonate (MMS) and temozolomide-induced DNA damages in T24 bladder carcinoma cells. 20 µM of Fen1i is shown to suppress the proliferation, migration, and invasion of tumor cells.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library