Skip to Content

SML4273

Nimustine hydrochloride

≥95% (HPLC)

Synonym(s):

N′-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-N-(2-chloroethyl)-N-nitroso-urea, 1-(4-Amino-2-methylpyrimidin-5-yl)methyl-3-(2-chloroethyl)-3-nitrosourea hydrochloride, Nimustine hydrochloride, ACNU hydrochloride, Nidran hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H13ClN6O2 · xHCl
Molecular Weight:
272.69 (free base basis)
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Nimustine HCl (ACNU) is a brain-penetrating nitrosourea that kills cancer cells by damaging their DNA. Nimustine HCl forms O6-chloroethylguanine adducts, which cross-link with other DNA strands, blocking replication and leading to cell death. Nimustine also suppresses blood cell production and is often used in combination with other chemotherapy drugs. Nimustine undergoes conversion into several active metabolites, including denitrosated ACNU, 3-((4-amino-2-methylpyrimidin-5-yl)methyl)-4-hydroxy-1-nitrosoimidazolidin-2-one, 1-(2-chloroethyl)-1-nitrosourea and 1-((4-amino-2-methylpyrimidin-5-yl)methyl)-5-hydroxyimidazolidin-2-one.

Disclaimer

Hygroscopic Store with desiccant


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Skin Sens. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library