Skip to Content

SML4277

FSEN1

≥98% (HPLC), powder, FSP1 inhibitor

Synonym(s):

3-(4-Bromophenyl)-6-[[4-(4-methoxyphenyl)-1-piperazinyl]methyl]thiazolo[2,3-c]-1,2,4-triazole, Ferroptosis sensitizer 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H22BrN5OS
CAS Number:
Molecular Weight:
484.41
MDL number:
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

FSEN1 (ferroptosis sensitizer 1) is a cell permeable, potent, and selective uncompetitive inhibitor of ferroptosis suppressor protein 1 (FSP1) that sensitizes cancer cells to ferroptosis. FSEN1 synergizes with inhibitors of GSH-GPX4 pathway and other reagents to induce ferroptosis.

FSEN1 binds to the substrate-binding pocket of FSP1, preventing it from catalyzing the reduction of coenzyme Q10 and other lipophilic quinones. This inhibition leads to an accumulation of lipid peroxides, promoting ferroptosis in cells that rely on FSP1 for survival. The selectivity of FSEN1 for human FSP1 over mouse FSP1 is attributed to specific interactions with a critical phenylalanine residue.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library