Skip to Content

SML4279

Validamycin A

≥95% (HPLC), powder, R. solani trehalase inhibitor

Synonym(s):

1,5,6-Trideoxy-4-O-β-D-glucopyranosyl-5-(hydroxymethyl)-1-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)-2-cyclohexen-1-yl]amino]-D-chiro-inositol, Validamycin, Validacin, Jinganmycin A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H35NO13
CAS Number:
Molecular Weight:
497.49
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Streptomyces hygroscopicus

Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

General description

Validamycin A (VMA) is a fungicidal antibiotic produced by Streptomyces hygroscopicus. It is an amino glycoside compound characterized by a sugar moiety without an amino group.

Application

Validamycin A may be used for evaluating its anti-seizure effectiveness in zebrafish larval epilepsy model; and studying its potential to induce plant defense responses and controlling fungal diseases.

Biochem/physiol Actions

Validamycin A is a potent and competitive inhibitor of Rhizoctonia solani trehalase. Validamycin A is used to protect rice plants against sheath blight caused by R.solani. Validamycin A in combination with amphotericin B appears to be effective against Aspergillus flavus strains resistant to amphotericin B. It is also efficient in controlling bacterial wilt intomatoes caused by P. solanacearum.


Still not finding the right product?

Explore all of our products under Validamycin A


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library